3,6-Dibromo-9-(3,4-dimethylphenyl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAD5420-1GEA
252
EUR
TCIAD5420-1G
3,6-Dibromo-9-(3,4-dimethylphenyl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
3,6-Dibromo-9-(3,4-dimethylphenyl)-9H-carbazole
Kaava:
C₂₀H₁₅Br₂N MW: 429,16 g/mol |
CAS-numero:
1786404-06-5 |
Testitulosten erittely
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 178.0 to 182.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...