Decanal Diethyl Acetal ≥98.0% (by GC)

Toimittaja: TCI EUROPE

TCIAD5755-5MLEA 39.6 EUR
TCIAD5755-5ML
Decanal Diethyl Acetal ≥98.0% (by GC)
Decanal Diethyl Acetal
Kaava: C₁₄H₃₀O₂
MW: 230,39 g/mol
Kiehumispiste: 92°C
MDL numero: MFCD00672804
CAS-numero: 34764-02-8
EINECS: 252-202-5

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR