Ethyl(3-methoxypropyl)dimethylammonium bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAE1282-5GEA 224 EUR
TCIAE1282-5G
Ethyl(3-methoxypropyl)dimethylammonium bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)
Ethyl(3-methoxypropyl)dimethylammonium bis(trifluoromethanesulphonyl)imide
Kaava: C₁₀H₂₀F₆N₂O₅S₂
MW: 426,39 g/mol
CAS-numero: 1373334-05-4

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid to cloudy liquid
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR