Fluorescein isothiocyanate dextran, M.W. ∼ 10000

Toimittaja: TCI EUROPE

TCIAF0918-100MGEA 117 EUR
TCIAF0918-100MG
Fluorescein isothiocyanate dextran, M.W. ∼ 10000
Fluorescein isothiocyanate dextran
Storage Temperature: Jääkaappi
MDL numero: MFCD00131092
CAS-numero: 60842-46-8

Order Now

Testitulosten erittely

Appearance Light yellow to Yellow to Orange powder to crystaline
Drying loss max. 7.0 %
Functionality test to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR