4-Cyano-3-fluorobenzaldehyde ≥97.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAF1112-1GEA 167 EUR
TCIAF1112-1G
4-Cyano-3-fluorobenzaldehyde ≥97.0% (by GC)
4-Cyano-3-fluorobenzaldehyde
Kaava: C₈H₄FNO
MDL numero: MFCD06738989
CAS-numero: 101048-76-4

Order Now

Testitulosten erittely

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 87.0 to 92.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR