N-Glycyl-DL-2-aminobutyric acid

Toimittaja: TCI
TCIAG0114-1GEA 53.9 EUR
TCIAG0114-1G
N-Glycyl-DL-2-aminobutyric acid
Proteiinit
  • Melting point: 230 °C
  • Solubility in wtaer: almost transparency
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR