N-Glycolyl-D-mannosamine Pentaacetate ≥93.0% (by HPLC)

Toimittaja: TCI EUROPE

TCIAG0463-100MGEA 327 EUR
TCIAG0463-100MG
N-Glycolyl-D-mannosamine Pentaacetate ≥93.0% (by HPLC)
N-Glycolyl-D-mannosamine Pentaacetate
Kaava: C₁₈H₂₅NO₁₂
MW: 447,39 g/mol
Storage Temperature: Jääkaappi
CAS-numero: 258824-38-3

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 93.0 area%
Elemental analysis (Nitrogen) 2.80 to 3.40 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR