(1H,1H,2H,2H-Heptadecafluorodecyl)phosphonic acid ≥96.0% (by HPLC, titration analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAH1459-200MGEA 57.2 EUR
TCIAH1459-200MG
(1H,1H,2H,2H-Heptadecafluorodecyl)phosphonic acid ≥96.0% (by HPLC, titration analysis)
(1H,1H,2H,2H-Heptadecafluorodecyl)phosphonic acid
Kaava: C₁₀H₆F₁₇O₃P
MW: 528,10 g/mol
MDL numero: MFCD19105612
CAS-numero: 80220-63-9

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %
Melting point 181.0 to 185.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR