2-(4-Hydroxyphenylazo)benzoic acid ≥98.0% (by HPLC, titration analysis) matrix for MALDI-TOF/MS

Toimittaja: TCI EUROPE
Varoitus

TCIAH1622-1GEA 62.7 EUR
TCIAH1622-1G
2-(4-Hydroxyphenylazo)benzoic acid ≥98.0% (by HPLC, titration analysis) matrix for MALDI-TOF/MS
2-(4-Hydroxyphenylazo)benzoic acid
Kaava: C₁₃H₁₀N₂O₃
MW: 242,23 g/mol
MDL numero: MFCD00002428
CAS-numero: 1634-82-8
EINECS: 216-655-2

Order Now

Testitulosten erittely

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR