Isomaltopentaose ≥95.0% (by HPLC)

Toimittaja: TCI EUROPE

TCIAI0854-50MGEA 168 EUR
TCIAI0854-50MG
Isomaltopentaose ≥95.0% (by HPLC)
Isomaltopentaose
Kaava: C₃₀H₅₂O₂₆
Storage Temperature: Pakastin
MDL numero: MFCD13182451
CAS-numero: 6082-32-2

Order Now

Testitulosten erittely

Purity (HPLC) min. 95.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR