Isomaltotetraose ≥95.0% (by HPLC)

Toimittaja: TCI EUROPE

TCIAI0855-50MGEA 130 EUR
TCIAI0855-50MG
Isomaltotetraose ≥95.0% (by HPLC)
Isomaltotetraose
Kaava: C₂₄H₄₂O₂₁
Storage Temperature: Pakastin
MDL numero: MFCD00133423
CAS-numero: 35997-20-7

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR