3-(Isopropoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Toimittaja: TCI EUROPE
Varoitus

TCIAI1000-1GEA 93.5 EUR
TCIAI1000-1G
3-(Isopropoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
3-(Isopropoxycarbonyl)phenylboronic acid
Kaava: C₁₀H₁₃BO₄
MW: 208,02 g/mol
MDL numero: MFCD02093047
CAS-numero: 342002-80-6

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR