Isopropylboronic acid pinacol ester ≥95.0% (by GC, titration analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAI1109-1GEA 86.9 EUR
TCIAI1109-1G
Isopropylboronic acid pinacol ester ≥95.0% (by GC, titration analysis)
Isopropylboronic acid pinacol ester
Kaava: C₉H₁₉BO₂
MW: 170,06
MDL numero: MFCD05663856
CAS-numero: 76347-13-2
UN: 3272
ADR: 3,III

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR