Loxoprofen ≥97.0% (by HPLC, titration analysis)

Toimittaja: TCI EUROPE

TCIAL0244-5GEA 116 EUR
TCIAL0244-5G
Loxoprofen ≥97.0% (by HPLC, titration analysis)
Loxoprofen
Kaava: C₁₅H₁₈O₃
MW: 246,31 g/mol
Sulamispiste: 111°C
MDL numero: MFCD00864331
CAS-numero: 68767-14-6

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR