4-Methoxypyridine-N-oxide ≥95.0% (by GC, titration analysis)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAM2401-5GEA
66
EUR
TCIAM2401-5G
4-Methoxypyridine-N-oxide ≥95.0% (by GC, titration analysis)
4-Methoxypyridine-N-oxide
Kaava:
C₆H₇NO₂ MW: 125,13 g/mol |
MDL numero:
MFCD00006207 CAS-numero: 1122-96-9 EINECS: 214-367-1 |
Testitulosten erittely
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...