[(4-Methoxyphenyl)ethynyl]trimethylsilane ≥97.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAM2785-1GEA 45.1 EUR
TCIAM2785-1G
[(4-Methoxyphenyl)ethynyl]trimethylsilane ≥97.0% (by GC)
[(4-Methoxyphenyl)ethynyl]trimethylsilane
Kaava: C₁₂H₁₆OSi
MW: 204,34 g/mol
Kiehumispiste: 96°C
MDL numero: MFCD04039887
CAS-numero: 3989-14-8

Order Now

Testitulosten erittely

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR