4'-Methoxy-3'-nitroacetophenone ≥98.0% (by GC)

Toimittaja: TCI EUROPE

TCIAM2811-5GEA 52.8 EUR
TCIAM2811-5G
4'-Methoxy-3'-nitroacetophenone ≥98.0% (by GC)
4'-Methoxy-3'-nitroacetophenone
Kaava: C₉H₉NO₄
MW: 195,18
MDL numero: MFCD00017023
CAS-numero: 6277-38-9
EINECS: 228-476-7

Order Now

Testitulosten erittely

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 100.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR