6-Methoxy-2-naphthalenemethanol ≥98.0% (by GC)

Toimittaja: TCI EUROPE

TCIAM2931-5GEA 251 EUR
TCIAM2931-5G TCIAM2931-1G
6-Methoxy-2-naphthalenemethanol ≥98.0% (by GC)
6-Methoxy-2-naphthalenemethanol
Kaava: C₁₂H₁₂O₂
MW: 188,23 g/mol
MDL numero: MFCD06202657
CAS-numero: 60201-22-1

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 120.0 to 124.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR