3-Methyl-5-isoxazolecarboxylic acid ≥98.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAM2953-1GEA 203 EUR
TCIAM2953-1G
3-Methyl-5-isoxazolecarboxylic acid ≥98.0% (by GC)
3-Methyl-5-isoxazolecarboxylic acid
Kaava: C₅H₅NO₃
MW: 127,10 g/mol
MDL numero: MFCD00464222
CAS-numero: 4857-42-5
EINECS: 225-454-9

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR