Diamond nanoparticles (Amine-modified) (<10 nm)
Toimittaja: TCI EUROPE
TCIAN0968-1GEA
149
EUR
TCIAN0968-1G
Diamond nanoparticles (Amine-modified) (<10 nm)
Diamond
Nanodiamonds (Nanocarbon Materials)
MDL numero:
MFCD00211867 CAS-numero: 7782-40-3 |
Testitulosten erittely
Appearance | White to Gray powder to crystal |
Purity | min. 97.0 % |
Water | max. 4.0 % |
Mean particle diameter | max. 10 nm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...