6-Nitro-2H-1,4-benzoxazin-3(4H)-one ≥98.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAN0999-1GEA 165 EUR
TCIAN0999-1G
6-Nitro-2H-1,4-benzoxazin-3(4H)-one ≥98.0% (by GC)
6-Nitro-2H-1,4-benzoxazin-3(4H)-one
Kaava: C₈H₆N₂O₄
MW: 194,15 g/mol
MDL numero: MFCD03425744
CAS-numero: 81721-87-1

Order Now

Testitulosten erittely

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 236.0 to 240.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR