4-Nitro-1,2,3,6,7,8-hexahydropyrene ≥98.0% (by HPLC, total nitrogen)

Toimittaja: TCI EUROPE

TCIAN1061-1GEA 143 EUR
TCIAN1061-1G
4-Nitro-1,2,3,6,7,8-hexahydropyrene ≥98.0% (by HPLC, total nitrogen)
4-Nitro-1,2,3,6,7,8-hexahydropyrene
Kaava: C₁₆H₁₅NO₂
MW: 253,30 g/mol
MDL numero: MFCD27964325
CAS-numero: 88535-47-1

Order Now

Testitulosten erittely

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 89.0 to 93.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR