3-Oxocyclopentanecarboxylic acid ≥98.0% (by GC, titration analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAO0416-1GEA 119 EUR
TCIAO0416-1G
3-Oxocyclopentanecarboxylic acid ≥98.0% (by GC, titration analysis)
3-Oxocyclopentanecarboxylic acid
Kaava: C6H8O3
MW: 128,13 g/mol
Kiehumispiste: 150°C
MDL numero: MFCD01320173
CAS-numero: 98-78-2

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 60.0 to 64.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR