Paclitaxel ≥98.0% (by HPLC)

Toimittaja: TCI EUROPE
Vaara

TCIAP1632-100MGEA 84.7 EUR
TCIAP1632-100MG
Paclitaxel ≥98.0% (by HPLC)
Paclitaxel
Kaava: C₄₇H₅₁NO₁₄
MDL numero: MFCD00869953
CAS-numero: 33069-62-4

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -49.0 to -54.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR