(R)-4-Phenylthiazolidine-2-thione ≥98.0% (by GC)

Toimittaja: TCI EUROPE

TCIAP1959-1GEA 153 EUR
TCIAP1959-1G
(R)-4-Phenylthiazolidine-2-thione ≥98.0% (by GC)
(R)-4-Phenylthiazolidine-2-thione
Kaava: C₉H₉NS₂
MDL numero: MFCD06658215
CAS-numero: 110199-18-3

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 128.0 to 130.0 deg-C
Specific rotation [a]20/D -204.0 to -207.0 deg (C=0.35, CHCL3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR