Pyrimidine-4-carboxylic acid ≥98.0% (by GC, titration analysis)

Toimittaja: TCI EUROPE

TCIAP1993-200MGEA 110 EUR
TCIAP1993-200MG
Pyrimidine-4-carboxylic acid ≥98.0% (by GC, titration analysis)
Pyrimidine-4-carboxylic acid
Kaava: C5H4N2O2
MW: 124,10 g/mol
MDL numero: MFCD00129737
CAS-numero: 31462-59-6
EINECS: 250-641-7

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 225.0 to 229.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR