1-Phenylcyclopentanecarbonitrile ≥98.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAP2146-1GEA
53.9
EUR
TCIAP2146-1G
TCIAP2146-5G
1-Phenylcyclopentanecarbonitrile ≥98.0% (by GC)
1-Phenylcyclopentanecarbonitrile
Kaava:
C₁₂H₁₃N MW: 171,24 g/mol Kiehumispiste: 108°C |
MDL numero:
MFCD00001358 CAS-numero: 77-57-6 EINECS: 201-038-2 |
Testitulosten erittely
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...