2-Phenylisonicotinic acid ≥98.0% (by GC, titration analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAP2453-1GEA 178 EUR
TCIAP2453-1G
2-Phenylisonicotinic acid ≥98.0% (by GC, titration analysis)
2-Phenylisonicotinic acid
Kaava: C₁₂H₉NO₂
MW: 199,2100 g/mol
MDL numero: MFCD03086008
CAS-numero: 55240-51-2

Order Now

Testitulosten erittely

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR