1-Phenyl-1H-indole ≥98.0% (by total nitrogen basis)

Toimittaja: TCI EUROPE
Varoitus

TCIAP2503-1GEA 141 EUR
TCIAP2503-1G
1-Phenyl-1H-indole ≥98.0% (by total nitrogen basis)
1-Phenyl-1H-indole
Kaava: C₁₄H₁₁N
MW: 193,25 g/mol
Kiehumispiste: 190°C
MDL numero: MFCD00123280
CAS-numero: 16096-33-6

Order Now

Testitulosten erittely

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR