Poly(2-methyl-2-oxazoline) n=∼100, ULTROXA®
Toimittaja: TCI EUROPE
TCIAP2506-200MGEA
88
EUR
TCIAP2506-200MG
Poly(2-methyl-2-oxazoline) n=∼100, ULTROXA®
Poly(2-methyl-2-oxazoline)
Storage Temperature:
Jääkaappi |
CAS-numero:
161358-46-9 |
Testitulosten erittely
Appearance | White to Light yellow to Light orange powder to crystal |
Average molecular weight | 8,000 to 12,250 |
Molecular weight distribution | max. 1.20 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...