Potassium bis(trimethylsilyl)amide 14% ∼0.6 mol/L in toluene

Toimittaja: TCI EUROPE
Vaara

TCIAP2730-100MLEA 67.1 EUR
TCIAP2730-100ML
Potassium bis(trimethylsilyl)amide 14% ∼0.6 mol/L in toluene
Potassium bis(trimethylsilyl)amide
Kaava: C₆H₁₈KNSi₂
MDL numero: MFCD00010330
CAS-numero: 40949-94-8
EINECS: 424-100-2
UN: 2924
ADR: 3,II

Order Now

Testitulosten erittely

Appearance Colorless to Light orange to Yellow clear liquid to cloudy liquid
Concentration (Neutralization titration) 12.0 to 14.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR