Rhodamine 590 chloride ≥90.0% (by HPLC)

Toimittaja: TCI EUROPE
Vaara

TCIAR0187-5GEA 168 EUR
TCIAR0187-5G
Rhodamine 590 chloride ≥90.0% (by HPLC)
Rhodamine 590 chloride
Kaava: C₂₇H₂₉ClN₂O₃
MW: 464,99 g/mol
MDL numero: MFCD01114540
CAS-numero: 3068-39-1
EINECS: 221-326-1
UN: 2811
ADR: 6.1,II

Order Now

Testitulosten erittely

Appearance Orange to Brown to Dark purple powder to crystal
Purity (HPLC) min. 90.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Lambda max. 524.0 to 528.0 nm (2mg/L H2O)
Absorbance (E1%1cm) min. 1,800 (2mg/L H2O, 524.0 to 528.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR