Rhodamine 800 ≥98.0% (by HPLC)

Toimittaja: TCI EUROPE
Varoitus

TCIAR0190-200MGEA 127 EUR
TCIAR0190-200MG
Rhodamine 800 ≥98.0% (by HPLC)
Rhodamine 800
Kaava: C₂₆H₂₆ClN₃O₅
MW: 495,96 g/mol
MDL numero: MFCD00467020
CAS-numero: 137993-41-0
UN: 1479
ADR: 5.1,III

Order Now

Testitulosten erittely

Appearance Green to Dark green to Dark blue powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Lambda max. 677.0 to 681.0 nm (MeOH)
Absorbance (E1%1cm) min. 2000 (MeOH, 677 to 681 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR