4,5,6-Trichloropyrimidine ≥98.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAT2550-1GEA 71.5 EUR
TCIAT2550-1G
4,5,6-Trichloropyrimidine ≥98.0% (by GC)
4,5,6-Trichloropyrimidine
Kaava: C₄HCl₃N₂
MW: 183,4231 g/mol
Kiehumispiste: 87°C
MDL numero: MFCD08458457
CAS-numero: 1780-27-4

Order Now

Testitulosten erittely

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR