4-(Trifluoromethyl)-1H-imidazole ≥98.0% (by GC, titration analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAT2928-1GEA 165 EUR
TCIAT2928-1G
4-(Trifluoromethyl)-1H-imidazole ≥98.0% (by GC, titration analysis)
4-(Trifluoromethyl)-1H-imidazole
Kaava: C₄H₃F₃N₂
MW: 136,08 g/mol
MDL numero: MFCD08458860
CAS-numero: 33468-69-8

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 149.0 to 153.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR