Tris(2-cyanoethyl) borate ≥98.0% (by titrimetric analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAT3024-5GEA 162 EUR
TCIAT3024-5G
Tris(2-cyanoethyl) borate ≥98.0% (by titrimetric analysis)
Tris(2-cyanoethyl) borate
Kaava: C₉H₁₂BN₃O₃
MW: 221,02 g/mol
CAS-numero: 126755-67-7

Order Now

Testitulosten erittely

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR