1,3,5-Trichloro-2,4,6-triiodobenzene ≥96.0% (by GC)

Toimittaja: TCI EUROPE

TCIAT3584-1GEA 164 EUR
TCIAT3584-1G
1,3,5-Trichloro-2,4,6-triiodobenzene ≥96.0% (by GC)
1,3,5-Trichloro-2,4,6-triiodobenzene
Kaava: C₆Cl₃I₃
MW: 559,13 g/mol
MDL numero: MFCD23135489
CAS-numero: 151721-79-8

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR