Label printers, M611
Toimittaja: Brady
116-0821EA 989 EUR
116-0852 116-0822 116-0821
Label printers, M611
Tulostimet ja tulostimien lisävarusteet Label Printers
The M611 Mobile Printer is a smartphone operated printer for cables and components, laboratory samples and complex labelling, electrical and datacom identification using the Brady Workstation Mobile Express Labels app.
  • Auto cutter
  • Monocolour
  • Barcode capability through software
You can easily create 1-D and 2-D codes and barcodes with a smartphone, serialise, use an extensive image library, include text, a time stamp and even data from the cloud in a complex label that can easily be designed in just a couple of steps. The Express app offers the most comprehensive label design capabilities available for any smartphone, and includes design wizards to quickly create specific identification labels.
Applications include asset identification; cable and wire identification; circuit board and component identification; datacom and audio, visual equipment labelling; electrical identification; general and industrial identification; laboratory identification; panel identification; rating and nameplate identification.
Materials supported include freezerbondz polyester; heat-shrink polyolefin sleeve; high temperature heat-shrink polyvinylidene fluoride sleeve; metallised polyester; metallised polyester with polyester overlaminate; nylon cloth; paper; paper with polyester overlaminate; polyester; polyester tag; polyethylene foam laminate polyester; polyethylene tag; polyimide; polypropylene; polypropylene tag; polyvinyl fluoride; rigid insert; self-laminating polyester; self-laminating vinyl; static dissipative polyester; tamper-evident metallised polyester; tamper-evident polyester; tamper-resistant metallised vinyl; tamper-resistant vinyl; thermoplastic polyether polyurethane; vinyl; vinyl cloth; vinyl tag.
Languages include Bulgarian; Croatian; Czech; Danish; Dutch; English; Estonian; finish; French; German; Hungarian; Italian; Norwegian (Bokmål); Polish; Portuguese; Romanian; Russian; Slovak; Slovenian; Spanish; Swedish; Turkish.
Toimitustiedot: This printer ships in a cardboard box holding a USB cable, a quick start guide, an AC powercord, a USB stick holding product info and Windows driver, a strap, either a Brady Workstation Basic Suite or a Brady Workstation Laboratory Identification Suite, a roll of PTL-97-488 and an M61-R4310 ribbon.