Diisopropyl disulphide 96%

Toimittaja: Thermo Scientific
Vaara

ACRO122700250EA 42 EUR
ACRO122700250
Diisopropyl disulphide 96%
Diisopropyl disulphide
Kaava: C₆H₁₄S₂
MW: 150,31 g/mol
Kiehumispiste: 175 °C (1013 hPa)
Sulamispiste: –69 °C
Tiheys: 0,943 g/cm³
Leimahduspiste: 18 °C
MDL numero: MFCD00008894
CAS-numero: 4253-89-8
EINECS: 224-225-0
UN: 1993

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4890 to 1.4930 (20°C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR