Cover glasses, Menzel Gläser
Toimittaja: Epredia
MENZDA020026A3E0EA 0 EUR
MENZDA020026A3E0 MENZDA020026A4E0_U
Cover glasses, Menzel Gläser
These cover slips are designed for use with hæmocytometers.
Pakkaus: Two pieces per plastic envelop - Ten pieces per box.