Labels, engraved plate replacement for M611 and BMP61 printer, B-7593
Toimittaja: Brady
BRDY197140EA 448 EUR
BRDY197140
Labels, engraved plate replacement for M611 and BMP61 printer, B-7593
Polyethylene foam laminate polyester engraved plate replacement labels for the BMP61 portable printer.
  • Gloss finish
  • Fan-folded layout
  • With stands −40 to 100 °C
Used for data and telecommunications labelling, electrical labelling, general identification.