Nitrile gloves, Amadex® Atlantis
Toimittaja: Amadex
Amadex® Atlantis
112-6057EA 20.1 EUR
112-6057 112-6058 112-6059 112-6060
Nitrile gloves, Amadex® Atlantis
Käsineet
Atlantis nitrile gloves are powder-free, online single chlorinated and non sterile.
  • Ambidextrous, finger textured, beaded cuff
  • 100% synthetic nitrile
  • No powder lubricant added
  • Shelf life of 5 years from the date of manufacture
Intended use: Non invasive devices for conducting medical examination, dentistry, clinical and laboratory applications.
Sertifikaatit: Conforms to ASTM D6319, EN 455, EN 420:2003+A1:2009, EN ISO 374-1:2016+A1:2018, EN 374-4:2019, EN 16523-1, EN 374-2, and EN ISO 374-5:2016; AQL 1,5.
Pakkaus: 100 gloves per box.