Nylon cloth labels for M611 and BMP™61 printers, type B-499
Toimittaja: Brady
BRDY150887EA 115 EUR
BRDY150887 BRDY150888 BRDY150889 BRDY150894 BRDY150895 BRDY150884 BRDY150885 BRDY150886 BRDY150890 BRDY150891 BRDY150892 BRDY150893
Nylon cloth labels for M611 and BMP™61 printers, type B-499
Etiketit, tarrat
Die-cut labels and continuous tapes are made from nylon cloth, with a permanent acrylic adhesive and matte finish.
  • Die-cut labels are supplied on a roll
  • Continuous tape is not cut to length; it is cut by the printer
  • Label thickness is 0,165 mm
  • Maximum service temperature: 90 °C
  • Minimum service temperature: –40 °C
  • Ribbon series: R4300, R6400
Nylon cloth (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as vials.