Eusolex® T-S
1.05334.9020EA 0 EUR
1.05334.9020 1.05334.0250
Eusolex® T-S
Eusolex®
Inorganic UV Filter
Eusolex® T-S is a hydrophobic Titanium Dioxide based, mineral UV filter with a vegetable surface coating. Eusolex® T-S shows a broad-spectrum UVA/UVB absorption and is COSMOS approved for natural cosmetics use.
INCI name: Titanium Dioxide, Alumina, Stearic Acid