Granisetron hydrochloride ≥98%

Toimittaja: Thermo Scientific
Varoitus

ACRO462480010EA 222 EUR
ACRO462480010
Granisetron hydrochloride ≥98%
Granisetron hydrochloride

  • Bioactive

Kaava: C₁₈H₂₄N₄O
Sulamispiste: 284…290 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01747034
CAS-numero: 107007-99-8

Order Now

Testitulosten erittely

Appearance White to almost white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
Solubility (1 % in water) Clear colorless solution
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR