Almotriptan Malate

Toimittaja: Thermo Scientific

ACRO465402500EA 476 EUR
ACRO465402500
Almotriptan Malate
Almotriptan Malate

  • Bioactive

Kaava: C₁₇H₂₅N₃O₂S·C₄H₆O₅
MW: 469,55 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD08067740
CAS-numero: 181183-52-8

Order Now

Testitulosten erittely

Appearance to light orange Powder
Infrared spectrum Conforms
Water ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR