Etoricoxib

Toimittaja: Thermo Scientific
Vaara

ACRO463380010EA 138 EUR
ACRO463380010
Etoricoxib
Etoricoxib

  • Bioactive

Kaava: C₁₈H₁₅ClN₂O₂S 
Sulamispiste: 134…138 °C
Storage Temperature: Pakastin
CAS-numero: 202409-33-4 
UN: 2811
ADR: 6.1,II

Order Now

Testitulosten erittely

Appearance White to light yellow Crsytalline powder
Infrared spectrum Conforms
Water ≤0.5 %
HPLC ≥97.5 %
Residue after ignition ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR