Benzophenone azine
Toimittaja: Thermo Scientific
ACRO464580010EA
148
EUR
ACRO464580010
Benzophenone azine
Benzophenone azine
- Bioactive
Kaava:
C₂₆H₂₀N₂ MW: 360,46 g/mol Sulamispiste: 161…162 °C Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00022006 CAS-numero: 983-79-9 |
Testitulosten erittely
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 160 to 165 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...