PBDB-T (PCE12), Organic Photovoltaics (OPV) reagent
Toimittaja: Thermo Scientific
H66526.MCEA
1230
EUR
H66526.MC
H66526.ME
H66526.MD
PBDB-T (PCE12), Organic Photovoltaics (OPV) reagent
PBDB-T (PCE12)
Kaava:
(C₆₈H₇₈O₂S₈)n |
CAS-numero:
1415929-80-4 |
Testitulosten erittely
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1,82 ± 0,02 eV |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...