VWR®, Centrifuge Tubes, High Speed, with Screw Cap
Toimittaja: VWR, part of Avantor
525-1590EA 284 EUR
VWRI525-1590
VWR®, Centrifuge Tubes, High Speed, with Screw Cap
Made from borosilicate glass 3.3. When used with rubber adapter sleeve they can withstand forces up to 13000×g.
  • Produced from borosilicate glass 3.3 tubing
  • Highly resistant against chemicals and rapid temperature changes
  • Autoclavable
  • With thread size 20-400, screw caps included
The VWR® high speed centrifuge tubes have been chemically strengthened to achieve a greater mechanical strength than standard test tubes.